siRNA Details
| Gene Symbol: Gm13288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243167.1 | NP_001230096.1 | 1 | 2360 | ||
| GenBank mRNA | AK031317.1 | - | 1 | 2361 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13288| Gene Symbol: Gm13288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243167.1 | NP_001230096.1 | 1 | 2360 | ||
| GenBank mRNA | AK031317.1 | - | 1 | 2361 | ||