siRNA Details
| Gene Symbol: Gm9047 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145360.1 | NP_001138832.1 | 1 | 496 | ||
| GenBank mRNA | BC147635.1 | - | 1 | 496 | 9056625 | |
| BC147645.1 | - | 1 | 496 | 9056635 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9047| Gene Symbol: Gm9047 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145360.1 | NP_001138832.1 | 1 | 496 | ||
| GenBank mRNA | BC147635.1 | - | 1 | 496 | 9056625 | |
| BC147645.1 | - | 1 | 496 | 9056635 | ||