siRNA Details
| Gene Symbol: Efr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082483.1 | NP_001075952.1 | 23 | 4362 | ||
| GenBank mRNA | AK129247.1 | - | 18 | 3677 | ||
| BC075650.1 | - | 22 | 4119 | |||
| BC147643.1 | - | 23 | 4362 | 9056633 | ||
| BC157968.1 | - | 23 | 4363 | 9088136 | ||

Efr3b
Genomic Map