siRNA Details
| Gene Symbol: Zfp936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034893.1 | NP_001030065.1 | 4 | 601 | ||
| GenBank mRNA | AK145615.1 | - | 4 | 601 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp936| Gene Symbol: Zfp936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034893.1 | NP_001030065.1 | 4 | 601 | ||
| GenBank mRNA | AK145615.1 | - | 4 | 601 | ||