siRNA Details
| Gene Symbol: Ssxb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205108.1 | NP_001192037.1 | 398 | |||
| GenBank mRNA | AY244797.1 | - | 399 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ssxb6| Gene Symbol: Ssxb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205108.1 | NP_001192037.1 | 398 | |||
| GenBank mRNA | AY244797.1 | - | 399 | |||