siRNA Details
| Gene Symbol: Myeov2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163425.1 | NP_001156897.1 | 3 | 323 | ||
| GenBank mRNA | AA764377.1 | - | 140 | |||
| AK003315.1 | - | 3 | 304 | |||
| AK012198.1 | - | 3 | 296 | |||
| AK152318.1 | - | 3 | 290 | |||
| BC049759.1 | - | 3 | 323 | |||
| BY631936.1 | - | 3 | 318 | |||

Myeov2
Genomic Map