siRNA Details
Gene Symbol: Rasgrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019211.2 | NP_062084.2 | 2 | 222 | ||
GenBank mRNA | AF081196.1 | - | 2 | 222 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rasgrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019211.2 | NP_062084.2 | 2 | 222 | ||
GenBank mRNA | AF081196.1 | - | 2 | 222 |