siRNA Details
| Gene Symbol: Vma21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081356.3 | NP_001074825.1 | 3 | 2913 | ||
| NM_001290780.1 | NP_001277709.1 | 3 | 3040 | |||
| NM_001290781.1 | NP_001277710.1 | 4 | 3214 | |||
| GenBank mRNA | AK164163.1 | - | 3 | 2868 | ||
| CX564875.1 | - | 1 | 112 | |||

Vma21
Genomic Map