siRNA Details
| Gene Symbol: Gm11567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101613.1 | NP_001095083.1 | 1 | 842 | ||
| GenBank mRNA | AK003994.1 | - | 1 | 843 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11567| Gene Symbol: Gm11567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101613.1 | NP_001095083.1 | 1 | 842 | ||
| GenBank mRNA | AK003994.1 | - | 1 | 843 | ||