siRNA Details
| Gene Symbol: Gm11565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126323.1 | NP_001119795.1 | 1 | 770 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11565| Gene Symbol: Gm11565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126323.1 | NP_001119795.1 | 1 | 770 | ||