siRNA Details
| Gene Symbol: Slc25a53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082412.2 | NP_001075881.1 | 5 | 514 | ||
| GenBank mRNA | AK012963.1 | - | 5 | 514 | ||
| AK013345.1 | - | 5 | 542 | |||
| AK028496.1 | - | 5 | 506 | |||
| AK049939.1 | - | 1 | 18 | |||
| AK076279.1 | - | 5 | 509 | |||
| BC024604.1 | - | 5 | 522 | |||

Slc25a53
Genomic Map