siRNA Details
Gene Symbol: Slc26a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019214.1 | NP_062087.1 | 2 | 272 | ||
XM_008764576.1 | XP_008762798.1 | 3 | 1596 | |||
GenBank mRNA | AF167412.1 | - | 2 | 272 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc26a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019214.1 | NP_062087.1 | 2 | 272 | ||
XM_008764576.1 | XP_008762798.1 | 3 | 1596 | |||
GenBank mRNA | AF167412.1 | - | 2 | 272 |