siRNA Details
| Gene Symbol: Gm9513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128510.1 | NP_001121982.1 | 2 | 163 | ||
| GenBank mRNA | EU703629.1 | - | 2 | 163 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9513| Gene Symbol: Gm9513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128510.1 | NP_001121982.1 | 2 | 163 | ||
| GenBank mRNA | EU703629.1 | - | 2 | 163 | ||