siRNA Details
| Gene Symbol: Herc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025992.2 | NP_080268.1 | 4, 5 | 700 | ||
| GenBank mRNA | AK011047.1 | - | 4, 5 | 587 | ||
| AK017888.1 | - | 4, 5 | 528 | |||
| AK147960.1 | - | 4, 5 | 624 | |||
| AK149648.1 | - | 4, 5 | 692 | |||

Herc6
Genomic Map