siRNA Details
| Gene Symbol: Parp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163575.1 | NP_001157047.1 | 2 | 175 | ||
| NM_001163576.1 | NP_001157048.1 | 1 | 279 | |||
| GenBank mRNA | AK028373.1 | - | 2 | 123 | ||
| AK035309.1 | - | 2 | 169 | |||
| AK046755.1 | - | 1 | 260 | |||
| AK146336.1 | - | 2 | 176 | |||
| AK153205.1 | - | 1 | 16 | |||
| AK166291.1 | - | 2 | 126 | |||
| BB843953.1 | - | 1 | 280 | |||
| BC024074.1 | - | 2 | 132 | |||
| BY742887.1 | - | 2 | 176 | |||

Parp10
Genomic Map