siRNA Details
| Gene Symbol: Rnf213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077880.1 | - | 7 | 1155 | ||
| AK150249.1 | - | 2 | 354 | |||
| AK156571.1 | - | 8 | 1288 | |||
| AK173199.1 | - | 17 | 4501 | |||
| BC022698.1 | - | 1 | 246 | |||
| BC038025.1 | - | 9 | 1446 | |||
| BC119284.1 | - | 6 | 976 | |||

Rnf213
Genomic Map