siRNA Details
| Gene Symbol: Rnf213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077880.1 | - | 1 | 100 | ||
| AK156571.1 | - | 2 | 233 | |||
| AK173199.1 | - | 11 | 3446 | |||
| BC038025.1 | - | 3 | 391 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf213| Gene Symbol: Rnf213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077880.1 | - | 1 | 100 | ||
| AK156571.1 | - | 2 | 233 | |||
| AK173199.1 | - | 11 | 3446 | |||
| BC038025.1 | - | 3 | 391 | |||