siRNA Details
| Gene Symbol: Parp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205239.1 | NP_001192168.1 | 2, 3 | 373 | ||
| NM_029922.3 | NP_084198.2 | 2, 3 | 373 | |||
| GenBank mRNA | AK007206.1 | - | 2, 3 | 297 | ||
| AK035349.1 | - | 2, 3 | 297 | |||
| AK076321.1 | - | 2, 3 | 297 | |||
| BC031387.1 | - | 1, 2 | 44 | 5038718 | ||
| BC052064.1 | - | 2, 3 | 371 | |||
| BC062096.1 | - | 1, 2 | 230 | 5716648 | ||
| BQ442721.1 | - | 2, 3 | 373 | |||

Parp6
Genomic Map