siRNA Details
| Gene Symbol: Zfand4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290338.1 | NP_001277267.1 | 6 | 1644 | ||
| NM_001290339.1 | NP_001277268.1 | 4 | 1344 | |||
| GenBank mRNA | AK006693.1 | - | 1 | 251 | ||
| AK012639.1 | - | 4 | 1343 | |||
| AK089032.1 | - | 6 | 1641 | |||

Zfand4
Genomic Map