siRNA Details
| Gene Symbol: Gm12171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163353.1 | NP_001156825.1 | 2 | 69 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12171| Gene Symbol: Gm12171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163353.1 | NP_001156825.1 | 2 | 69 | ||