siRNA Details
| Gene Symbol: 4930453N24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026273.2 | NP_080549.1 | 3 | 1313 | ||
| GenBank mRNA | AK002578.1 | - | 3 | 1308 | ||
| AK075826.1 | - | 3 | 1297 | |||
| AK161311.1 | - | 3 | 1314 | |||
| BC020029.1 | - | 3 | 1302 | 4012943 | ||

4930453N24Rik
Genomic Map