siRNA Details
| Gene Symbol: 4930523C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162896.1 | NP_001156368.1 | 3 | 985 | ||
| GenBank mRNA | AK028922.1 | - | 3 | 788 | ||
| AK034159.1 | - | 3 | 681 | |||
| AK035814.1 | - | 3 | 980 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930523C07Rik| Gene Symbol: 4930523C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162896.1 | NP_001156368.1 | 3 | 985 | ||
| GenBank mRNA | AK028922.1 | - | 3 | 788 | ||
| AK034159.1 | - | 3 | 681 | |||
| AK035814.1 | - | 3 | 980 | |||