siRNA Details
| Gene Symbol: 4930523C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162896.1 | NP_001156368.1 | 3 | 3026 | ||
| GenBank mRNA | AK028660.1 | - | 1065 | |||
| AK028922.1 | - | 3 | 2829 | |||
| AK054156.1 | - | 623 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930523C07Rik| Gene Symbol: 4930523C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162896.1 | NP_001156368.1 | 3 | 3026 | ||
| GenBank mRNA | AK028660.1 | - | 1065 | |||
| AK028922.1 | - | 3 | 2829 | |||
| AK054156.1 | - | 623 | ||||