siRNA Details
| Gene Symbol: Cyp4f37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100187.1 | NP_001093657.1 | 13 | 2578 | ||
| GenBank mRNA | AK132492.1 | - | 13 | 2578 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp4f37| Gene Symbol: Cyp4f37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100187.1 | NP_001093657.1 | 13 | 2578 | ||
| GenBank mRNA | AK132492.1 | - | 13 | 2578 | ||