siRNA Details
| Gene Symbol: Fcrl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164725.1 | NP_001158197.1 | 5 | 785 | ||
| GenBank mRNA | DQ470838.1 | - | 5 | 790 | ||
| DQ470839.1 | - | 4 | 1086 | |||
| DQ470840.1 | - | 5 | 781 | |||
| EF032497.1 | - | 5 | 694 | |||

Fcrl6
Genomic Map