siRNA Details
| Gene Symbol: Zmym4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114399.1 | NP_001107871.1 | 30 | 5270 | ||
| GenBank mRNA | AK039026.1 | - | 1 | 356 | ||
| BC029670.1 | - | 3 | 1108 | |||
| BC038024.1 | - | 8 | 1719 | |||
| BC050924.1 | - | 24 | 4179 | |||

Zmym4
Genomic Map