siRNA Details
| Gene Symbol: Zmym4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114399.1 | NP_001107871.1 | 14 | 2478 | ||
| GenBank mRNA | AK129137.1 | - | 14 | 2733 | ||
| BC050924.1 | - | 8 | 1387 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zmym4| Gene Symbol: Zmym4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114399.1 | NP_001107871.1 | 14 | 2478 | ||
| GenBank mRNA | AK129137.1 | - | 14 | 2733 | ||
| BC050924.1 | - | 8 | 1387 | |||