siRNA Details
| Gene Symbol: Camsap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081360.1 | NP_001074829.1 | 13 | 1971 | ||
| GenBank mRNA | AK028542.1 | - | 11 | 1921 | ||
| BC138367.1 | - | 12 | 1547 | 8861387 | ||
| BC145258.1 | - | 11 | 1514 | 9053811 | ||

Camsap2
Genomic Map