siRNA Details
| Gene Symbol: Pdzd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040136.2 | NP_001035226.1 | 4 | 1215 | ||
| NM_001040137.2 | NP_001035227.1 | 4 | 985 | |||
| GenBank mRNA | AK005921.1 | - | 4 | 1216 | ||
| AK006727.1 | - | 4 | 1119 | |||
| AK015115.1 | - | 4 | 986 | |||
| AK017019.1 | - | 4 | 1123 | |||
| BC147551.1 | - | 4 | 1211 | 9056542 | ||
| BC147563.1 | - | 4 | 1211 | 9056553 | ||
| BC172089.1 | - | 3 | 1021 | |||

Pdzd9
Genomic Map