siRNA Details
| Gene Symbol: 9430016H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081181.2 | NP_001074650.1 | 1 | 652 | ||
| GenBank mRNA | AK007831.1 | - | 1 | 441 | ||
| AK079116.1 | - | 1 | 654 | |||
| AK165130.1 | - | 1 | 444 | |||
| BC024945.1 | - | 1 | 121 | 3989472 | ||
| BC071241.1 | - | 449 | 5125554 | |||

9430016H08Rik
Genomic Map