siRNA Details
| Gene Symbol: 9430016H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081181.2 | NP_001074650.1 | 5 | 1281 | ||
| GenBank mRNA | AK079116.1 | - | 5 | 1283 | ||
| BC071241.1 | - | 1078 | 5125554 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9430016H08Rik| Gene Symbol: 9430016H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081181.2 | NP_001074650.1 | 5 | 1281 | ||
| GenBank mRNA | AK079116.1 | - | 5 | 1283 | ||
| BC071241.1 | - | 1078 | 5125554 | |||