siRNA Details
| Gene Symbol: AI846148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033139.3 | NP_001028311.2 | 6 | 2473 | ||
| GenBank mRNA | AK090385.1 | - | 6 | 2443 | ||
| AK142757.1 | - | 5 | 1870 | |||
| BC125249.1 | - | 6 | 2301 | 40059530 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI846148| Gene Symbol: AI846148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033139.3 | NP_001028311.2 | 6 | 2473 | ||
| GenBank mRNA | AK090385.1 | - | 6 | 2443 | ||
| AK142757.1 | - | 5 | 1870 | |||
| BC125249.1 | - | 6 | 2301 | 40059530 | ||