siRNA Details
| Gene Symbol: AI846148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033139.3 | NP_001028311.2 | 6 | 3119 | ||
| GenBank mRNA | AK090385.1 | - | 6 | 3089 | ||
| AK142757.1 | - | 5 | 2516 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI846148| Gene Symbol: AI846148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033139.3 | NP_001028311.2 | 6 | 3119 | ||
| GenBank mRNA | AK090385.1 | - | 6 | 3089 | ||
| AK142757.1 | - | 5 | 2516 | |||