siRNA Details
| Gene Symbol: AI846148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167831.1 | NP_001161303.1 | 5 | 1455 | ||
| GenBank mRNA | AK168010.1 | - | 5 | 1349 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI846148| Gene Symbol: AI846148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167831.1 | NP_001161303.1 | 5 | 1455 | ||
| GenBank mRNA | AK168010.1 | - | 5 | 1349 | ||