siRNA Details
| Gene Symbol: Sgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193568.1 | NP_001180497.1 | 2 | 251 | ||
| NM_001193569.1 | NP_001180498.1 | 2 | 189 | |||
| NM_019232.3 | NP_062105.2 | 2 | 163 | |||
| XM_006227723.1 | XP_006227785.1 | 4 | 849 | |||
| XM_008758606.1 | XP_008756828.1 | 2 | 359 | |||
| GenBank mRNA | BC078843.1 | - | 2 | 117 | 7130800 | |
| CB814192.1 | - | 2 | 189 | |||
| FM042519.1 | - | 2 | 251 | |||
| FM068981.1 | - | 2 | 169 | |||
| FM123924.1 | - | 2 | 156 | |||
| L01624.1 | - | 2 | 168 | |||

Sgk1
Genomic Map