siRNA Details
| Gene Symbol: 0610030E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026696.1 | NP_080972.1 | 4 | 2472 | ||
| GenBank mRNA | AK085662.1 | - | 1 | 3683 | ||
| BC066053.1 | - | 4 | 2446 | 6827710 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

0610030E20Rik| Gene Symbol: 0610030E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026696.1 | NP_080972.1 | 4 | 2472 | ||
| GenBank mRNA | AK085662.1 | - | 1 | 3683 | ||
| BC066053.1 | - | 4 | 2446 | 6827710 | ||