siRNA Details
| Gene Symbol: Ptx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163416.1 | NP_001156888.1 | 2, 3 | 790 | ||
| NM_026747.1 | NP_081023.1 | 2, 3 | 880 | |||
| GenBank mRNA | AK003777.1 | - | 1, 2 | 349 | ||
| BC107380.2 | - | 2, 3 | 773 | 40057084 | ||
| BC107381.2 | - | 2, 3 | 863 | 40057088 | ||
| BC118508.1 | - | 2, 3 | 863 | 40106397 | ||

Ptx4
Genomic Map