siRNA Details
| Gene Symbol: Dcdc2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177964.2 | NP_001171435.2 | 8 | 719 | ||
| GenBank mRNA | AK003729.1 | - | 8 | 720 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcdc2c| Gene Symbol: Dcdc2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177964.2 | NP_001171435.2 | 8 | 719 | ||
| GenBank mRNA | AK003729.1 | - | 8 | 720 | ||