siRNA Details
| Gene Symbol: Eml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001043335.1 | NP_001036800.1 | 22 | 3588 | ||
| NM_001043336.2 | NP_001036801.1 | 22 | 3205 | |||
| NM_001286346.1 | NP_001273275.1 | 22 | 3342 | |||
| NM_001286347.1 | NP_001273276.1 | 23 | 3256 | |||
| GenBank mRNA | AK003593.1 | - | 1 | 633 | ||
| AK132592.1 | - | 3 | 1106 | |||
| BC038273.1 | - | 10 | 1853 | |||
| BC053094.1 | - | 22 | 3422 | 6400458 | ||
| BC059839.1 | - | 22 | 3183 | 6417403 | ||
| BC079582.1 | - | 22 | 3241 | 5702743 | ||
| BC098329.1 | - | 20 | 3623 | |||

Eml1
Genomic Map