siRNA Details
| Gene Symbol: Gpr155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190297.2 | NP_001177226.1 | 17 | 3457 | ||
| NM_001276443.1 | NP_001263372.1 | 17 | 3510 | |||
| NM_001276444.1 | NP_001263373.1 | 16 | 3373 | |||
| GenBank mRNA | BC026382.1 | - | 7 | 1473 | ||
| HM161763.2 | - | 17 | 3434 | |||
| HM161764.2 | - | 17 | 3487 | |||
| HM161766.2 | - | 16 | 3350 | |||

Gpr155
Genomic Map