siRNA Details
| Gene Symbol: Smim6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162998.1 | NP_001156470.1 | 2 | 953 | ||
| GenBank mRNA | AK003745.1 | - | 2 | 953 | ||
| AK156823.1 | - | 2 | 1086 | |||
| BC038330.1 | - | 2 | 631 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim6| Gene Symbol: Smim6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162998.1 | NP_001156470.1 | 2 | 953 | ||
| GenBank mRNA | AK003745.1 | - | 2 | 953 | ||
| AK156823.1 | - | 2 | 1086 | |||
| BC038330.1 | - | 2 | 631 | |||