siRNA Details
| Gene Symbol: Ccdc167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163741.2 | NP_001157213.1 | 1 | 43 | ||
| NM_026782.2 | NP_081058.1 | 1 | 43 | |||
| GenBank mRNA | AK003900.1 | - | 1 | 38 | ||
| AK049389.1 | - | 12 | ||||
| AK085526.1 | - | 12 | ||||
| BC022730.1 | - | 1 | 20 | |||
| BY067347.1 | - | 1 | 45 | |||

Ccdc167
Genomic Map