siRNA Details
| Gene Symbol: Ccdc167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163741.2 | NP_001157213.1 | 5 | 3646 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc167| Gene Symbol: Ccdc167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163741.2 | NP_001157213.1 | 5 | 3646 | ||