siRNA Details
| Gene Symbol: Fndc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081416.1 | NP_001074885.1 | 18 | 4924 | ||
| GenBank mRNA | AK003938.1 | - | 1 | 45 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fndc1| Gene Symbol: Fndc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081416.1 | NP_001074885.1 | 18 | 4924 | ||
| GenBank mRNA | AK003938.1 | - | 1 | 45 | ||