siRNA Details
| Gene Symbol: Klk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026806.2 | NP_081082.1 | 5 | 1212 | ||
| GenBank mRNA | AK003996.1 | - | 5 | 1213 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klk5| Gene Symbol: Klk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026806.2 | NP_081082.1 | 5 | 1212 | ||
| GenBank mRNA | AK003996.1 | - | 5 | 1213 | ||