siRNA Details
| Gene Symbol: Klk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026806.2 | NP_081082.1 | 1 | 22 | ||
| GenBank mRNA | AK003996.1 | - | 1 | 23 | ||
| BC139475.1 | - | 1 | 23 | 8862497 | ||
| BC139478.1 | - | 1 | 23 | 8862500 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klk5| Gene Symbol: Klk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026806.2 | NP_081082.1 | 1 | 22 | ||
| GenBank mRNA | AK003996.1 | - | 1 | 23 | ||
| BC139475.1 | - | 1 | 23 | 8862497 | ||
| BC139478.1 | - | 1 | 23 | 8862500 | ||