siRNA Details
| Gene Symbol: Fam172a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163419.1 | NP_001156891.1 | 10 | 1682 | ||
| NM_001163420.1 | NP_001156892.1 | 9 | 1490 | |||
| NM_138312.1 | NP_612185.1 | 11 | 1806 | |||
| GenBank mRNA | AK052419.1 | - | 10 | 1655 | ||
| AK165273.1 | - | 11 | 1705 | |||
| AK166285.1 | - | 9 | 1463 | |||
| BC057380.1 | - | 10 | 1650 | |||

Fam172a
Genomic Map