siRNA Details
| Gene Symbol: Fam172a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163419.1 | NP_001156891.1 | 2 | 250 | ||
| NM_001163420.1 | NP_001156892.1 | 2 | 250 | |||
| NM_138312.1 | NP_612185.1 | 3 | 374 | |||
| GenBank mRNA | AF064782.1 | - | 2 | 203 | ||
| AK049727.1 | - | 2 | 211 | |||
| AK052419.1 | - | 2 | 222 | |||
| AK165273.1 | - | 2 | 224 | |||
| AK166285.1 | - | 2 | 222 | |||
| AK199247.1 | - | 1 | 45 | |||
| BC057380.1 | - | 2 | 218 | |||

Fam172a
Genomic Map