siRNA Details
| Gene Symbol: Rnf166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033142.2 | NP_001028314.1 | 6 | 1742 | ||
| GenBank mRNA | AK004000.1 | - | 1 | 369 | ||
| AK079883.1 | - | 6 | 1729 | |||
| AK151813.1 | - | 6 | 1742 | |||
| AK193864.1 | - | 1 | 132 | |||
| AK209147.1 | - | 1 | 132 | |||

Rnf166
Genomic Map