siRNA Details
| Gene Symbol: Flnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081185.1 | NP_001074654.1 | 46 | 8030 | ||
| GenBank mRNA | AF119148.1 | - | 1 | 122 | ||
| AK004264.1 | - | 1 | 123 | |||
| AK135523.1 | - | 11 | 1870 | |||
| BC039885.1 | - | 11 | 1898 | |||
| BC052186.1 | - | 19 | 2979 | |||
| BC060276.1 | - | 25 | 4091 | |||
| BC151097.1 | - | 46 | 7835 | 9088010 | ||
| BC158128.1 | - | 45 | 7736 | 9007396 | ||
| BC172159.1 | - | 45 | 7736 | |||

Flnc
Genomic Map